Ethyl-2-amino-5-methylbenzoate
Supplier: Apollo Scientific
Synonyms: Ethyl 2-amino-5-methylbenzoate
OR110167-1G OR110167-5G
APOSOR110167-1GEA 0 EUR
APOSOR110167-1G APOSOR110167-5G
Ethyl-2-amino-5-methylbenzoate
Ethyl 2-amino-5-methylbenzoate
Formula: C₁₀H₁₃NO₂
MW: 179.2157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00115547
CAS Number: 58677-05-7