2-(1H-Pyrazol-3-yl)-1,3-thiazole
Supplier: Apollo Scientific
Warning
OR110205-1G OR110205-250MG
APOSOR110205-1GEA 132 EUR
APOSOR110205-1G APOSOR110205-250MG
2-(1H-Pyrazol-3-yl)-1,3-thiazole
2-(1H-Pyrazol-3-yl)-1,3-thiazole
Formula: C₆H₅N₃S
MW: 151.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00105072
CAS Number: 166196-73-2