3-Bromo-6-chloroimidazo[1,2-a]pyridine
Supplier: Apollo Scientific
Warning
OR110278-1G OR110278-5G
APOSOR110278-1GEA 277 EUR
APOSOR110278-1G APOSOR110278-5G
3-Bromo-6-chloroimidazo[1,2-a]pyridine
3-Bromo-6-chloroimidazo[1,2-a]pyridine
Formula: C₇H₄BrClN₂
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD08667858
CAS Number: 886371-28-4