3-Bromoimidazo[1,2-a]pyridine-6-carbaldehyde
Supplier: Apollo Scientific
Warning
OR110284-1G OR110284-250MG
APOSOR110284-1GEA 474 EUR
APOSOR110284-1G APOSOR110284-250MG
3-Bromoimidazo[1,2-a]pyridine-6-carbaldehyde
3-Bromoimidazo[1,2-a]pyridine-6-carbaldehyde
Formula: C₈H₅BrN₂O
MW: 225.05 g/mol
Melting Pt: 207…210 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD14583145
CAS Number: 1004550-20-2