Ethyl (6-chloroimidazo[1,2-a]pyrid-2-yl)acetate
Supplier: Apollo Scientific
OR110350-1G OR110350-5G
APOSOR110350-1GEA 134 EUR
APOSOR110350-1G APOSOR110350-5G
Ethyl (6-chloroimidazo[1,2-a]pyrid-2-yl)acetate
Ethyl (6-chloroimidazo[1,2-a]pyrid-2-yl)acetate
Formula: C₁₁H₁₁ClN₂O₂
MW: 238.68 g/mol
Melting Pt: 56…59 °C
Storage Temperature: Ambient
MDL Number: MFCD10699212
CAS Number: 59128-02-8