4-Bromo-3,5-dihydroxybenzoic acid 98%
Supplier: Apollo Scientific
Warning
OR11038-100G OR11038-500G OR11038-250G
APOSOR11038-100GEA 161 EUR
APOSOR11038-100G APOSOR11038-500G APOSOR11038-250G
4-Bromo-3,5-dihydroxybenzoic acid 98%
4-Bromo-3,5-dihydroxybenzoic acid
Formula: C₇H₅BrO₄
MW: 233.02 g/mol
Melting Pt: 268…276 °C
Storage Temperature: Ambient
MDL Number: MFCD00002513
CAS Number: 16534-12-6
EINECS: 240-606-4