Methyl-4-hydroxy-3-nitrobenzoate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 4-hydroxy-3-nitrobenzoate
OR110386-100G
APOSOR110386-100GEA 121 EUR
APOSOR110386-100G
Methyl-4-hydroxy-3-nitrobenzoate 98%
Methyl 4-hydroxy-3-nitrobenzoate
Formula: C₈H₇NO₅
MW: 197.15 g/mol
Melting Pt: 73…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00014695
CAS Number: 99-42-3