Ethyl pyrazine-2-carboxylate
Supplier: Apollo Scientific
OR110404-1G OR110404-5G OR110404-250MG
APOSOR110404-1GEA 37.7 EUR
APOSOR110404-1G APOSOR110404-5G APOSOR110404-250MG
Ethyl pyrazine-2-carboxylate
Ethyl pyrazine-2-carboxylate
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 120 °C/6mmHg
Melting Pt: 49…52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00792887
CAS Number: 6924-68-1