2-(4-Bromophenyl)-5-methyl-1,3,4-oxadiazole
Supplier: Apollo Scientific
Warning
OR110415-1G OR110415-10G OR110415-5G
APOSOR110415-1GEA 161 EUR
APOSOR110415-1G APOSOR110415-10G APOSOR110415-5G
2-(4-Bromophenyl)-5-methyl-1,3,4-oxadiazole
2-(4-Bromophenyl)-5-methyl-1,3,4-oxadiazole
Formula: C₉H₇BrN₂O
Melting Pt: 107…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00491620
CAS Number: 41421-03-8