Methyl-1H-pyrazole-3-carboxylate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 1H-pyrazole-3-carboxylate
OR110467-25G OR110467-10G
APOSOR110467-25GEA 31.9 EUR
APOSOR110467-25G APOSOR110467-10G
Methyl-1H-pyrazole-3-carboxylate 98%
Methyl-1H-pyrazole-3-carboxylate
Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
Melting Pt: 141…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00649381
CAS Number: 15366-34-4