4-(4-Bromophenoxy)benzaldehyde
Supplier: Apollo Scientific
Synonyms: 4-(4-Bromo-phenoxy)benzaldehyde
OR110490-1G OR110490-5G
APOSOR110490-1GEA 63.8 EUR
APOSOR110490-1G APOSOR110490-5G
4-(4-Bromophenoxy)benzaldehyde
4-(4-Bromophenoxy)benzaldehyde
Formula: C₁₃H₉BrO₂
MW: 277.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276811
CAS Number: 69240-56-8