3',4'-Dimethoxyacetophenone
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3,4-Dimethoxyphenyl)ethan-1-one, 3,4-Dimethoxyacetophenone, 1-(3,4-Dimethoxyphenyl)ethanone
OR11051-500G
APOSOR11051-500GEA 127 EUR
APOSOR11051-500G
3',4'-Dimethoxyacetophenone
3',4'-Dimethoxyacetophenone
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 286…288 °C
Melting Pt: 49…51 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00008737
CAS Number: 1131-62-0
EINECS: 214-468-0