3-Ethyl-4-methyl-3-pyrrolin-2-one
Supplier: Apollo Scientific
Warning
Synonyms: 3-Ethyl-4-methyl-2,5-dihydro-1H-pyrrol-2-one, 1,5-Dihydro-3-ethyl-4-methyl-2H-pyrrol-2-one
OR11055-100G
APOSOR11055-100GEA 79.9 EUR
APOSOR11055-100G
3-Ethyl-4-methyl-3-pyrrolin-2-one
3-Ethyl-4-methyl-3-pyrrolin-2-one
Formula: C₇H₁₁NO
MW: 125.17 g/mol
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00173861
CAS Number: 766-36-9