tert-Butyl-4-[2-(4-bromo-1H-pyrazol-1-yl)ethyl]piperazine-1-carboxylate
Supplier: Apollo Scientific
OR110576-1G OR110576-5G
APOSOR110576-1GEA 248 EUR
APOSOR110576-1G APOSOR110576-5G
tert-Butyl-4-[2-(4-bromo-1H-pyrazol-1-yl)ethyl]piperazine-1-carboxylate
tert-Butyl-4-[2-(4-bromo-1H-pyrazol-1-yl)ethyl]piperazine-1-carboxylate
Formula: C₁₄H₂₃BrN₄O₂
MW: 359.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100120
CAS Number: 877401-28-0