1-Tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-amine
Supplier: Apollo Scientific
Warning
Synonyms: 1-Tetrahydropyran-4-yl-1H-pyrazol-4-ylamine, 1-(Tetrahydro-2H-pyran-4-yl)-1H-pyrazol-4-amine
OR110577-1G OR110577-250MG OR110577-5G
APOSOR110577-1GEA 139 EUR
APOSOR110577-1G APOSOR110577-250MG APOSOR110577-5G
1-Tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-amine
1-Tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-amine
Formula: C₈H₁₃N₃O
MW: 167.21 g/mol
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD11505007
CAS Number: 1190380-49-4