5,6-Dihydro-4H-cyclopenta[b]thiophen-2-ylmethanol
Supplier: Apollo Scientific
Warning
OR110634-1G OR110634-5G
APOSOR110634-1GEA 296 EUR
APOSOR110634-1G APOSOR110634-5G
5,6-Dihydro-4H-cyclopenta[b]thiophen-2-ylmethanol
5,6-Dihydro-4H-cyclopenta[b]thiophen-2-ylmethanol
Formula: C₈H₁₀OS
MW: 154.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08059488
CAS Number: 41301-23-9