4-Methyl-1,3-thiazole-5-sulphonyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 4-Methylthiazole-5-sulphonyl chloride
OR110665-100MG OR110665-1G OR110665-250MG
APOSOR110665-100MGEA 40.6 EUR
APOSOR110665-100MG APOSOR110665-1G APOSOR110665-250MG
4-Methyl-1,3-thiazole-5-sulphonyl chloride
4-Methyl-1,3-thiazole-5-sulphonyl chloride
Formula: C₄H₄ClNO₂S₂
MW: 197.66 g/mol
Melting Pt: 34…36 °C
Storage Temperature: Freezer
MDL Number: MFCD09864968
CAS Number: 953070-51-4
UN: 3261
ADR: 8 III