4-Methylbenzophenone 99%
Supplier: Apollo Scientific
Warning
OR11067-1KG OR11067-250G OR11067-50G
APOSOR11067-1KGEA 74.1 EUR
APOSOR11067-1KG APOSOR11067-250G APOSOR11067-50G
4-Methylbenzophenone 99%
4-Methylbenzophenone
Formula: C₁₄H₁₂O
Boiling Pt: 326 °C
Melting Pt: 56.5…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00008553
CAS Number: 134-84-9