Methyl-1,5-dimethyl-1H-pyrazole-3-carboxylate
Supplier: Apollo Scientific
Synonyms: Methyl 1,5-dimethyl-1H-pyrazole-3-carboxylate, Methyl 1,5-Dimethylpyrazole-3-carboxylate, 1,5-Dimethylpyrazole-3-carboxylic acid methyll ester
OR110685-1G OR110685-5G
APOSOR110685-1GEA 0 EUR
APOSOR110685-1G APOSOR110685-5G
Methyl-1,5-dimethyl-1H-pyrazole-3-carboxylate
Methyl-1,5-dimethyl-1H-pyrazole-3-carboxylate
Formula: C₇H₁₀N₂O₂
MW: 154.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06661312
CAS Number: 10250-61-0