4,6-Dichloro-2H-thiochromene-3-carbaldehyde
Supplier: Apollo Scientific
OR110693-1G OR110693-5G
APOSOR110693-1GEA 60.9 EUR
APOSOR110693-1G APOSOR110693-5G
4,6-Dichloro-2H-thiochromene-3-carbaldehyde
4,6-Dichloro-2H-thiochromene-3-carbaldehyde
Formula: C₁₀H₆Cl₂OS
MW: 245.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00084993
CAS Number: 54949-15-4