2'-Hydroxyacetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: o-Hydroxyacetophenone, 2-Acetophenol, 2-Hydroxyphenyl Methyl Ketone, 1-(2-Hydroxyphenyl)ethan-1-one, o-Acetylphenol, 1-(2-Hydroxyphenyl)-1-oxoethane, 1-Ethanoyl-2-hydroxybenzol, 2-Acetylphenol
OR11070-1KG OR11070-500G
APOSOR11070-1KGEA 142 EUR
APOSOR11070-1KG APOSOR11070-500G
2'-Hydroxyacetophenone 98%
2'-Hydroxyacetophenone
Formula: C₈H₈O₂
Boiling Pt: 90…92 °C/10mmHg
Melting Pt: 44350 °C
Density: 1.131 g/cm³ (25 °C)
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00002219
CAS Number: 118-93-4
EINECS: 204-288-0