2-Chloro-5-nitroanisole
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-2-methoxy-4-nitrobenzene
OR11086-25G OR11086-5G
APOSOR11086-25GEA 26.1 EUR
APOSOR11086-25G APOSOR11086-5G
2-Chloro-5-nitroanisole
2-Chloro-5-nitroanisole
Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00079739
CAS Number: 1009-36-5
EINECS: 213-768-9