3-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid
Supplier: Apollo Scientific
OR110873-1G OR110873-5G
APOSOR110873-1GEA 69.6 EUR
APOSOR110873-1G APOSOR110873-5G
3-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid
3-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid
Formula: C₁₃H₁₅NO₃
MW: 233.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00664642
CAS Number: 101004-93-7