3-Bromo-o-xylene 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Bromo-2,3-dimethylbenzene
OR11088-100G
OR11088-25G
OR11088-500G
APOSOR11088-100GEA
116
EUR
APOSOR11088-100G
APOSOR11088-25G
APOSOR11088-500G
3-Bromo-o-xylene 99%
3-Bromo-o-xylene
Formula:
C₈H₉Br MW: 185.06 g/mol Boiling Pt: 212…214 °C Density: 1.365 g/cm³ (25 °C) Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000069 CAS Number: 576-23-8 EINECS: 209-398-2 |