6-Chloro-8-iodoimidazo[1,2-a]pyridine
Supplier: Apollo Scientific
OR110902-1G OR110902-5G
APOSOR110902-1GEA 65.4 EUR
APOSOR110902-1G APOSOR110902-5G
6-Chloro-8-iodoimidazo[1,2-a]pyridine
6-Chloro-8-iodoimidazo[1,2-a]pyridine
Formula: C₇H₄ClIN₂
Storage Temperature: Ambient
MDL Number: MFCD10758078
CAS Number: 1033463-28-3