2-Chloro-4-(1H-imidazol-1-yl)pyrimidine
Supplier: Apollo Scientific
OR110940-1G OR110940-250MG OR110940-500MG
APOSOR110940-1GEA 43.5 EUR
APOSOR110940-1G APOSOR110940-250MG APOSOR110940-500MG
2-Chloro-4-(1H-imidazol-1-yl)pyrimidine
2-Chloro-4-(1H-imidazol-1-yl)pyrimidine
Formula: C₇H₅ClN₄
MW: 180.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09756941
CAS Number: 114834-03-6