5-Methoxy[1,3]thiazolo[5,4-b]pyridin-2-amine
Supplier: Apollo Scientific
Warning
Synonyms: 5-Methoxythiazolo[5,4-b]pyridin-2-amine
OR110973-1G OR110973-250MG
APOSOR110973-1GEA 203 EUR
APOSOR110973-1G APOSOR110973-250MG
5-Methoxy[1,3]thiazolo[5,4-b]pyridin-2-amine
5-Methoxy[1,3]thiazolo[5,4-b]pyridin-2-amine
Formula: C₇H₇N₃OS
MW: 181.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00100555
CAS Number: 13797-77-8