1-(2,6-Dichlorophenyl)-2-thiourea
Supplier: Apollo Scientific
|
Danger
|
OR11103-100G
OR11103-25G
OR11103-5G
APOSOR11103-100GEA
174
EUR
APOSOR11103-100G
APOSOR11103-25G
APOSOR11103-5G
1-(2,6-Dichlorophenyl)-2-thiourea
1-(2,6-Dichlorophenyl)-2-thiourea
Formula:
(C₆H₃Cl₂)NC(S)NH₂ Melting Pt: 155…159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041156 CAS Number: 6590-91-6 UN: 2811 ADR: 6.1,III |