2-Acetyl-3,4-dibromothiophene
Supplier: Apollo Scientific
OR111072-10G OR111072-1G OR111072-5G
APOSOR111072-10GEA 852 EUR
APOSOR111072-10G APOSOR111072-1G APOSOR111072-5G
2-Acetyl-3,4-dibromothiophene
2-Acetyl-3,4-dibromothiophene
Formula: C₆H₄Br₂OS
Storage Temperature: Ambient
CAS Number: 57681-57-9