3-Bromo-o-toluidine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-6-bromotoluene, 3-Bromo-2-methylaniline
OR11112-100G OR11112-25G OR11112-5G
APOSOR11112-100GEA 121 EUR
APOSOR11112-100G APOSOR11112-25G APOSOR11112-5G
3-Bromo-o-toluidine 98%
3-Bromo-o-toluidine
Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 253…255 °C
Density: 1.511 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00051579
CAS Number: 55289-36-6
EINECS: 259-568-5