1-Methylpyrrole-2-carbonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 1-Methyl-1H-pyrrole-2-carbonitrile, 2-Cyano-1-methylpyrrole
OR111182-1G OR111182-5G
APOSOR111182-1GEA 55.1 EUR
APOSOR111182-1G APOSOR111182-5G
1-Methylpyrrole-2-carbonitrile
1-Methylpyrrole-2-carbonitrile
Formula: C₆H₆N₂
MW: 106.13 g/mol
Density: 1.03 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02693721
CAS Number: 34884-10-1