Biphenyl-4-sulphonyl chloride 95%
Supplier: Apollo Scientific
Danger
Synonyms: Biphenyl-4-sulphonyl Chloride
OR11127-25G OR11127-5G OR11127-10G
APOSOR11127-25GEA 29 EUR
APOSOR11127-25G APOSOR11127-5G APOSOR11127-10G
Biphenyl-4-sulphonyl chloride 95%
Biphenyl-4-sulphonyl chloride
Formula: C₁₂H₉ClO₂S
MW: 252.72 g/mol
Boiling Pt: 200…205 °C
Melting Pt: 112…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00078658
CAS Number: 1623-93-4
EINECS: 216-609-1
UN: 3261
ADR: 8 II