1-Bromo-2-methoxyethane 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromoethyl methyl ether
OR11137-100G OR11137-500G OR11137-1KG
APOSOR11137-100GEA 79.9 EUR
APOSOR11137-100G APOSOR11137-500G APOSOR11137-1KG
1-Bromo-2-methoxyethane 98%
1-Bromo-2-methoxyethane
Formula: BrH₂CCH₂OCH₃
MW: 138.99 g/mol
Boiling Pt: 109…110 °C
Density: 1.478 g/cm³ (25 °C)
Flash Pt: 28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000236
CAS Number: 6482-24-2
EINECS: 229-339-4
UN: 3271
ADR: 3 III