3,3-Dimethyl-1-butyne
Supplier: Apollo Scientific
Danger
Synonyms: 3,3-Dimethylbutyne
OR11141-25G OR11141-100G OR11141-500G
APOSOR11141-25GEA 284 EUR
APOSOR11141-25G APOSOR11141-100G APOSOR11141-500G
3,3-Dimethyl-1-butyne
3,3-Dimethyl-1-butyne
Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 37…38 °C
Melting Pt: –78 °C
Density: 0.671 g/cm³ (25 °C)
Flash Pt: –25 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008852
CAS Number: 917-92-0
EINECS: 213-035-3
UN: 3295
ADR: 3,II