4-Dimethoxymethylpyrimidin-2-ylamine
Supplier: Apollo Scientific
OR111450-1G OR111450-5G
APOSOR111450-1GEA 92.8 EUR
APOSOR111450-1G APOSOR111450-5G
4-Dimethoxymethylpyrimidin-2-ylamine
4-Dimethoxymethylpyrimidin-2-ylamine
Formula: C₇H₁₁N₃O₂
MW: 169.1800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05864792
CAS Number: 165807-05-6