5-Methoxybenzimidazole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Methoxy-1H-benzo[d]imidazole
OR11201-10G
OR11201-1G
OR11201-5G
APOSOR11201-10GEA
88.6
EUR
APOSOR11201-10G
APOSOR11201-1G
APOSOR11201-5G
5-Methoxybenzimidazole
5-Methoxybenzimidazole
Formula:
C₈H₈N₂O MW: 148.17 g/mol Melting Pt: 121…126 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00272526 CAS Number: 4887-80-3 |