6-Nitro-o-toluidine
Supplier: Apollo Scientific
Danger
Synonyms: 6-Nitro-2-methylaniline, 2-Amino-3-nitrotoluene, 2-Methyl-6-nitroaniline
OR11212-100G OR11212-10G OR11212-25G
APOSOR11212-100GEA 43.5 EUR
APOSOR11212-100G APOSOR11212-10G APOSOR11212-25G
6-Nitro-o-toluidine
6-Nitro-o-toluidine
Formula: C₇H₈N₂O₂
Melting Pt: 93…96 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007744
CAS Number: 570-24-1
EINECS: 209-329-6
UN: 2660
ADR: 6.1,III