4-Nitrophenyl chloroformate
Supplier: Apollo Scientific
Danger
OR11227-100G OR11227-25G OR11227-5G
APOSOR11227-100GEA 72.5 EUR
APOSOR11227-100G APOSOR11227-25G APOSOR11227-5G
4-Nitrophenyl chloroformate
4-Nitrophenyl chloroformate
Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Boiling Pt: 159…162 °C/19mmHg
Melting Pt: 77…79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007321
CAS Number: 7693-46-1
EINECS: 231-706-9
UN: 3261
ADR: 8 II