Bis(4-methoxyphenyl)disulphide 99%
Supplier: Apollo Scientific
Danger
Synonyms: 1,2-Bis(4-methoxyphenyl)disulphane
OR11235-1G OR11235-25G OR11235-5G
APOSOR11235-1GEA 21.9 EUR
APOSOR11235-1G APOSOR11235-25G APOSOR11235-5G
Bis(4-methoxyphenyl)disulphide 99%
Bis(4-methoxyphenyl) disulphide
Formula: C₁₄H₁₄O₂S₂
MW: 278.4 g/mol
Boiling Pt: 200 °C/3mmHg
Melting Pt: 40…45 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00041358
CAS Number: 5335-87-5
UN: 3077
ADR: 9+EHS III