6-Chloropurine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 6-Chloro-7H-purine, 6-Chloro-1H-purine, 6-Chloro-9H-purine
OR11255-100G OR11255-25G OR11255-500G
APOSOR11255-100GEA 34.8 EUR
APOSOR11255-100G APOSOR11255-25G APOSOR11255-500G
6-Chloropurine 97%
6-Chloropurine
Formula: C₅H₃ClN₄
MW: 154.56 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00075825
CAS Number: 87-42-3
EINECS: 201-745-6