ω-Bromo-2-hydroxyacetophenone
Supplier: Apollo Scientific
Danger
Synonyms: 2-Hydroxyphenacyl bromide, 2-Bromo-1-(2-hydroxyphenyl)ethan-1-one, 2-Bromo-1-(2-hydroxyphenyl)ethanone, α-Bromo-2-hydroxyacetophenone, 2-Bromo-2'-hydroxyacetophenone
OR11268-25G OR11268-5G OR11268-100G
APOSOR11268-25GEA 177 EUR
APOSOR11268-25G APOSOR11268-5G APOSOR11268-100G
ω-Bromo-2-hydroxyacetophenone
ω-Bromo-2-hydroxyacetophenone
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD01727570
CAS Number: 2491-36-3
UN: 3263
ADR: 8,II