2,6-Xylidine
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-m-xylene, 2,6-Dimethylaniline
OR11273-1KG OR11273-500G
APOSOR11273-1KGEA 87 EUR
APOSOR11273-1KG APOSOR11273-500G
2,6-Xylidine
2,6-Xylidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 214 °C/739mmHg
Melting Pt: 44540 °C
Density: 0.984 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007747
CAS Number: 87-62-7
EINECS: 201-758-7
UN: 1711
ADR: 6.1,II