3'-Hydroxyacetophenone
Supplier: Apollo Scientific
Warning
OR11287-100G OR11287-25G
APOSOR11287-100GEA 37.7 EUR
APOSOR11287-100G APOSOR11287-25G
3'-Hydroxyacetophenone
3'-Hydroxyacetophenone
Formula: C₈H₈O₂
MW: 136.15 g/mol
Melting Pt: 95…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00002298
CAS Number: 121-71-1
EINECS: 204-494-0