3',5'-Dimethoxyacetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3,5-Dimethoxyphenyl)ethan-1-one, 1-(3,5-Dimethoxyphenyl)ethanone
OR11298-1G OR11298-5G
APOSOR11298-1GEA 37.7 EUR
APOSOR11298-1G APOSOR11298-5G
3',5'-Dimethoxyacetophenone 98%
3',5'-Dimethoxyacetophenone
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 290…291 °C
Melting Pt: 33…41 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00008739
CAS Number: 39151-19-4
EINECS: 254-322-3