3,5-Dimethoxybenzyl bromide 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1-(Bromomethyl)-3,5-dimethoxybenzene
OR11305-1G OR11305-25G OR11305-5G
APOSOR11305-1GEA 21.9 EUR
APOSOR11305-1G APOSOR11305-25G APOSOR11305-5G
3,5-Dimethoxybenzyl bromide 97%
3,5-Dimethoxybenzyl bromide
Formula: C₉H₁₁BrO₂
MW: 231.0864 g/mol
Melting Pt: 69…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01321368
CAS Number: 877-88-3
UN: 3261
ADR: 8,II