2-Chloro-5-(chloromethyl)pyridine
Supplier: Apollo Scientific
Danger
OR11306-100G OR11306-25G OR11306-500G
APOSOR11306-100GEA 48 EUR
APOSOR11306-100G APOSOR11306-25G APOSOR11306-500G
2-Chloro-5-(chloromethyl)pyridine
2-Chloro-5-(chloromethyl)pyridine
Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Melting Pt: 37…42 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00125366
CAS Number: 70258-18-3
UN: 3261
ADR: 8,II