3-Methoxy-4-methylbenzaldehyde
Supplier: Apollo Scientific
Warning
OR11329-5G OR11329-10G OR11329-1G
APOSOR11329-5GEA 34.8 EUR
APOSOR11329-5G APOSOR11329-10G APOSOR11329-1G
3-Methoxy-4-methylbenzaldehyde
3-Methoxy-4-methylbenzaldehyde
Formula: C9H10O2
MW: 150.1700 g/mol
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD04114050
CAS Number: 24973-22-6