Benzoin methyl ether 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1,2-Diphenyl-2-methoxyethan-1-one, 2-Methoxy-1,2-diphenylethanone
OR11336-100G OR11336-25G
APOSOR11336-100GEA 116 EUR
APOSOR11336-100G APOSOR11336-25G
Benzoin methyl ether 98%
Benzoin methyl ether
Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Melting Pt: 47…50 °C
Density: 1.128 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008492
CAS Number: 3524-62-7
EINECS: 222-538-7
UN: 2811
ADR: 6.1 III