4-(4-Methoxyphenoxy)aniline 98+%
Supplier: Apollo Scientific
Danger
OR11346-5G OR11346-1G OR11346-10G
APOSOR11346-5GEA 24.8 EUR
APOSOR11346-5G APOSOR11346-1G APOSOR11346-10G
4-(4-Methoxyphenoxy)aniline 98+%
4-(4-Methoxyphenoxy)aniline
Formula: C₁₃H₁₃NO₂
MW: 215.2478 g/mol
Boiling Pt: 245…250 °C/33mmHg
Melting Pt: 75…79 °C
Density: 1.294 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044105
CAS Number: 31465-36-8
EINECS: 250-644-3