4-(2-Chlorophenoxy)benzenesulphonyl chloride 95%
Supplier: Apollo Scientific
Danger
OR11349-1G OR11349-5G
APOSOR11349-1GEA 127 EUR
APOSOR11349-1G APOSOR11349-5G
4-(2-Chlorophenoxy)benzenesulphonyl chloride 95%
4-(2-Chlorophenoxy)benzenesulphonyl chloride
Formula: C₁₂H₈Cl₂O₃S
Melting Pt: 89…94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631899
CAS Number: 610277-84-4
UN: 3261
ADR: 8,II